Principaux documents
CLS6795410D
Corning® stirrer with digital display
AC/DC input 120 V AC, US 3-pin plug, plate L × W 5 in. × 7 in.
Synonyme(s) :
magnetic stirrer, magnetic stirrers, stirrers
About This Item
Code UNSPSC :
41103803
Nomenclature NACRES :
NB.73
Conditionnement
case of 1
Fabricant/nom de marque
Corning 6795-410D
Disponibilité
not available in EU
Paramètres
60-1150 rpm speed
Plaque/Plateau, L × l
5 in. × 7 in.
Description générale
  • Digital LED stir speed display indicates when set speed has been reached
  • Microprocessor maintains consistent and repeatable stir speed settings
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
Autres remarques
individual units are not routinely calibrated, rpm speeds are approximate only
Informations légales
Corning is a registered trademark of Corning, Inc.