Principaux documents
CLS64062LFO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 1 ea
Synonyme(s) :
equipment replacement accessory
About This Item
Code UNSPSC :
41121809
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Corning 6406-2LFO
Capacité
2 L
Volume
2 L
20000 mL
Description générale
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Informations légales
PYREX is a registered trademark of Corning, Inc.