Principaux documents
CLS4985P1L
Corning® narrow mouth Erlenmeyer flask, reusable
capacity 1 L, polypropylene, with GL-52 PP screw cap
Synonyme(s) :
conical flasks, laboratory flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
narrow-mouth
polypropylene
screw top cap
screw top cap
Stérilité
non-sterile
Caractéristiques
cap
graduated 10 mL (intervals)
Conditionnement
pack of 4 ea
Fabricant/nom de marque
Corning 4985P-1L
Capacité
1 L
Description générale
Translucent polypropylene flasks are manufactured with blue graduations for reference. The flasks have a screw neck and are supplied with a polypropylene cap.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.