CLS4519174
Corning® ProCulture® spinner flask accessories
side arm fitting, Dual with combination 1/8 & 1/4 inlets for 500 mL spinner flask, Gas and Media Handling Fitting with Combination 1/8 in. & 1/4 in. Inlets for 500mL Angled Sidearm
Synonyme(s) :
container accessory
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.43
Produits recommandés
Fabricant/nom de marque
Corning 4519-174
Catégories apparentées
Description générale
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Informations légales
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries