Principaux documents
CLS3835
Corning® non-treated 1536 well microplates
black, clear bottom, low base, w/o lid, non-sterile
Synonyme(s) :
1536 multiwell plates, 1536 well microplates, 1536 well microtiter plates, 1536 well plates
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.13
Matériaux
black
Stérilité
non-sterile
Caractéristiques
black, clear bottom
Conditionnement
case of 100 ea
Fabricant/nom de marque
Corning 3835
Taille
1536 wells
Adéquation
suitable for (cell cuture)
Informations légales
Corning is a registered trademark of Corning, Inc.