Principaux documents
CLS2679940
Corning® polypropylene closure
fits neck O.D. 40 mm
, orange
About This Item
Code UNSPSC :
41121800
Matériaux
orange
polypropylene
Conditionnement
case of 100
Catégories apparentées
Description générale
For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.
Informations légales
Corning is a registered trademark of Corning, Inc.