Principaux documents
CLS1395OR
Corning® Silicone O-Ring
for use with GL45 Pyrex® Bottles
About This Item
Code UNSPSC :
41121815
Stérilité
non-sterile
Conditionnement
pack of 10
Description générale
These silicone O-rings are used in place of the pouring rings on PYREX GL 45 bottles to ensure a leak-resistant seal.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.