Principaux documents
C1420
Cuvette washer
Double
Synonyme(s) :
cuvette cleaning
About This Item
Code UNSPSC :
41121813
Nomenclature NACRES :
SB.32
Description générale
Supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvettes.