BR701377
BRAND® sealing film for microplates
suitable for PCR, storage, aluminum film, DMSO/solvent resistant, operating temperature (-80)—120 °C, pack of 100 ea
Synonyme(s) :
701377, BRAND film, PCR sealing film, aluminum film, cover, plate film, seal, sealing film, storage film
About This Item
Produits recommandés
Matériaux
aluminum film
Stérilité
non-sterile
Caractéristiques
DNA free
RNase and DNase free
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 701377
Paramètres
(-80)—120 °C operating temperature
Adéquation
suitable for PCR
Description générale
Caractéristiques et avantages
- Temperature stability to 120°C and down to -80°C
- Tight seal for minimizing evaporation
- DMSO- and solvent-resistant
- Removable without leaving residue