Principaux documents
BMSIPS205020
Sigma-Aldrich
Benchmark OS-20L Overhead Stirrer
AC/DC input 120 V
Synonyme(s) :
overhead electric stirrer, digital overhead stirrer
About This Item
Code UNSPSC :
41103800
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific IPS2050-20
Paramètres
30-2200 rpm speed
Description générale
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Caractéristiques et avantages
  • LCD control panel
  • Includes complete stand assembly
  • Compact and quiet
  • Digital accuracy and convenience