Principaux documents
BMSIPD960035PTFE
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Pharmaceutical Samples
PTFE, capacity 35 mL, pack of 1
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.82
Matériaux
PTFE
Conditionnement
pack of 1
Fabricant/nom de marque
Benchmark Scientific IPD9600-35PTFE
Capacité
35 mL
Compatibilité
BeadBlaster 96
Description générale
Samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. All of the jars feature threaded closures for leakproof performance with both solids and liquids. PTFE is the least hard of the materials used in the jars and is chosen for its inert properties.
PTFE jars are metal-free and non-stick. They are highly temperature resistant, expanding their use to cryogrinding. PTFE jars are used for applications in the pharmaceutical field as well as for plants and other biological samples. The 15mm PTFE grinding balls have steel cores for a higher energy input. The jars are supplied as a 35ml volume
Application
Homogenization of Pharmaceutical samples, cryogrinding
Informations légales
BeadBlaster is a trademark of Benchmark Scientific, Inc.