BAH378620152
Replacement Parts for Scienceware® Hand Held Colony Counter
Red Felt Tip Pen
About This Item
Code UNSPSC :
41104000
Nomenclature NACRES :
NB.22
Produits recommandés
Fabricant/nom de marque
Bel-Art H37862-0152
Description générale
- Works on plastic or glass
- Is oil and water resistant
- Is removable with ethyl alcohol
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP