AXYSCT150SSRS
Self standing screw cap tubes and attached caps with O-rings
capacity 0.5 mL, red, sterile
Synonyme(s) :
centrifuge tubes
About This Item
Matériaux
red
screw top cap
Stérilité
sterile
Caractéristiques
RCF 20,000 × g
Conditionnement
pack of 500 tubes
case of 8 packs
Fabricant/nom de marque
Axygen Inc
Corning
Capacité
0.5 mL