Principaux documents
AXYP384240SQC
Corning® Axygen® Deep Well Microplate
size 384 wells, Not Treated, working volume 240 μL, pkg of 10x5plates/cs
Synonyme(s) :
Deep well plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
square bottom clear polypropylene wells
Stérilité
non-sterile
Caractéristiques
lid: no
skirt
Not Treated
Fabricant/nom de marque
Axygen Inc P-384-240SQ-C
Corning
Conditionnement
pkg of 10x5plates/cs
Volume maximum
250 μL
Taille
384 wells
Volume de travail
240 μL
Adéquation
suitable for PCR
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.