Z531162
Aldrich® coiled condenser
unjacketed, L 400 mm, joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS24/40
Longueur
400 mm
Description générale
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC