Principaux documents
YY2103119-M
Millipore
Gauge 1/4 in. NPTM vacuum/pressure gauge 0-30 in. (760 mm) Hg vacuum  and 0-7 bar (0 100 psi)
About This Item
Code UNSPSC :
40161525
eCl@ss :
32031603
Matériaux
316L stainless steel
Niveau de qualité
D.I.
6.3 mm (0.25 in.)