Principaux documents
XX6370007
Millipore
UV lamp type G8T5
Beer, Bottled Water, Cider, Clinical Lab, Cosmetics, Environmental Analysis, Food and Beverage, Industrial QC, Laboratory Filtration, Microbial monitoring, Product Testing, Water Monitoring, Wine
About This Item
Code UNSPSC :
41104424
Niveau de qualité
Application(s)
cosmetics
environmental
food and beverages
water monitoring
Compatibilité
for use with Ultraviolet Sterilizer (XX6370000/05)
Description générale
Rapidly decontaminate 47 mm Millipore filter holders between successive sample filtrations
Clause de non-responsabilité
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.