Principaux documents
VPMH107000
Millipore
Viresolve® Pro Magnus Holder
Synonyme(s) :
Viresolve® Pro Magnus Holder (for 1 to 7 Devices)
About This Item
Code UNSPSC :
23151806
eCl@ss :
32031690
Matériaux
316L stainless steel component(s) (structural)
Niveau de qualité
Fabricant/nom de marque
Viresolve®
L × l × H
135 cm (53 in.) × 61 cm (24 in.) × 56 cm (22 in.)
Description générale
The Viresolve® Magnus Holders are designed for large volume processing. The Viresolve® Pro Magnus Holder is designed to run the Viresolve® Pro Device. No product contact eliminates the need for cleaning validations. This cartridge holder, VPMH107000, is capable of accepting 1 to 7 units
Caractéristiques et avantages
  • Simple manual hydraulics to clamp units
  • Vertical orientation to minimize footprint and facilitate use (stack, clamp, run)
Informations légales
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany