Principaux documents
MWTC
Millipore
mPAGE® Mini Wet Transfer Cassette
replacement part for mPAGE® Mini Wet Transfer System
About This Item
Code UNSPSC :
41105305
Nomenclature NACRES :
NA.32
Conditionnement
pkg of 1 pack
Fabricant/nom de marque
Millipore
Technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
Compatibilité
for use with mPAGE® Mini Wet Transfer System.
Description générale
The mPAGE® Mini Wet Transfer Cassette is a replacement part for the mPAGE® Mini Wet Transfer System (MWTS).
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Informations légales
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany