Principaux documents
MA0HC10FS1
Millipore
Filtre en profondeur Millistak+® HC, format Pod
(A0HC media series), cellulose membrane (and inorganic filter aid), effective filtration area 1.1 m2
About This Item
Code UNSPSC :
23151806
Matériaux
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
Niveau de qualité
Mode de stérilisation compatible
autoclavable compatible
Surface de filtration efficace
1.1 m2
Raccords
seal (flat seal)
Code de la cartouche
(A0HC media series)
Description générale
Millistak+® A0HC media series Depth Filters is offered in a scalable, disposable format. Accommodating applications from lab to pilot to process scale, the Process-Scale Pod format offers greater flexibility because of its unique modular design.
Millistak+® Pod system is ideal for a wide variety of Primary and Secondary Clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
The Millistak+® Pod Filters are available in three distinct series of media grades in order to meet your specific application needs. The Millistak+® A0HC media series delivers optimal performance through a gradient density matrix as well as positive surface charge properties.
Informations légales
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany