Principaux documents
1.01111
Supelco
Reducer (stainless steel)
for 2" to S40 thread
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NA.03
Raccords
for 2" to S40 thread
Température de stockage
no temp limit
Description générale
This reducer connects our barrels and drums with G2 opening to S40-threaded equipment. It is equipped with a G2 outer thread and an S40 outer thread.
from 2" to S40 thread