Accéder au contenu
MilliporeSigma
Toutes les photos(1)

Principaux documents

03-0193-00

Millipore

SMC® Human SARS-CoV-2 RBD IgG Kit

for protein sequence analysis, for protein analysis

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Code UNSPSC :
12161503
eCl@ss :
32161000
Nomenclature NACRES :
NA.84

Qualité

for protein analysis

Technique(s)

single molecule counting: suitable

Description générale

The assay variation of the SMC Human SARS-CoV-2 RBD IgG Immunoassay kit were studied using twenty-one plasma samples, with roughly half positive and half negative, run in triplicate by 2 different operators on 2 different days. Mean intra-assay variation for all samples was < 15%. Mean inter-assay variation for all samples was < 21%.

Application

This new streamline kit has the following features including:

  • For use with the state of the art TM=["SMCxPRO"] Ultrasensitive Immunoassay System


  • Qualitative assay; for research use only<br />• Detects SARS-CoV-2 RBD specific IgG antibody within 3 days of PCR positive result


  • Requires as little as 2?L of human serum and plasma sample (1:50 recommended dilution)


  • Fast assay protocol (one hour capture, 30 minute detection steps)

Composants

Assay Buffer SARS-CoV-2 RBD IgG Beads Standard Diluent SARS-CoV-2 RBD IgG Detection Antibody 10X Wash Buffer Buffer D Elution Buffer B

Informations légales

SMC is a registered trademark of Singulex

Clause de non-responsabilité

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictogrammes

Health hazardExclamation mark

Mention d'avertissement

Danger

Mentions de danger

Classification des risques

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Code de la classe de stockage

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificats d'analyse (COA)

Recherchez un Certificats d'analyse (COA) en saisissant le numéro de lot du produit. Les numéros de lot figurent sur l'étiquette du produit après les mots "Lot" ou "Batch".

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique