Principaux documents
Z743296
Julabo® adaptors
twin distributing, one m16x1 female to 2 x m16x1 male, pack of 1
Synonyme(s) :
barbed adaptor
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Conditionnement
pack of 1
Fabricant/nom de marque
Julabo 8970473
Description générale
For use with Julabo® circulators with M16x1 female fittings, twin distributing adapter with M16x1 female to 2 x M16x1 male. Pack of 1.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.