Principaux documents
Z742524
Block for digital dry bath and MyBlock
Holds 12 x 15-16 mm tubes
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.22
Caractéristiques
Holds 12 x 15-16 mm tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific BSW1516
Catégories apparentées
Description générale
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Informations légales
MyBlock is a trademark of Benchmark Scientific, Inc.