Principaux documents
Z742367
2MAG MIXcontrol MTP control device
AC/DC input 100-240 V, US 3-pin plug
Synonyme(s) :
control unit, power bank, power unit
About This Item
Code UNSPSC :
41103800
Nomenclature NACRES :
NB.72
Matériaux
stainless steel housing
stainless steel plate
Caractéristiques
4 step power setting (25/50/75/100%)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
2mag 90250
Paramètres
0-40 °C temp. limit
100-2000 rpm speed
Larg. × P × H
200 mm × 155 mm × 38 mm
Taille de la plaque, larg. × Long.
× 200 mm × 120 mm, no of position= 12
Poids
~1.4 kg
Description générale
Universal control unit for the connection of one stirring drive MIXdrive MTP, universal 10 watts stirring power, wide speed range from 100 up to 2,000 rpm, jerk-free stirring also at low speeds, 4-step power setting.
Clear digital display for speed- and power settings, SoftStart for reliable centering and safe accelerating of the stirring bar.