Principaux documents
Z740723
Sigma-Aldrich
Aldrich® Essentials boiling flask, flat-bottom, ST/NS 24/40
capacity 250 mL
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass flask
flat bottom
single-neck
Conditionnement
pkg of 1 ea
Capacité
250 mL
Catégories apparentées
Description générale
Clear borosilicate glass, flat bottomed boiling flasks with top, outer ST ground joint neck. White marking spot and capacity.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC