Principaux documents
Z740644
Sigma-Aldrich
Aldrich® Essentials desiccator
I.D. × H 210 mm × 310 mm, with perforated porcelain plate
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
D.I. × H
210 mm × 310 mm
Description générale
Heavy-wall, molded, neutral glass, desiccators with ground flange top and lid. Lid has knob top to assist in removing lid. Complete with perforated, bottom, porcelain plate. No direct heat or flame can be used.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC