Principaux documents
Z693790
LabJaws lattice rod S-connector
nickel plated
Synonyme(s) :
clamp holders, frame connectors
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Matériaux
nickel plated
Fabricant/nom de marque
Troemner 916127
Capacité de la mâchoire
0-13 mm
Description générale
Clamps two lattice rods at a 90° angle and fetures separate adjustment screws for each rod location.
Informations légales
LabJaws is a trademark of Henry Troemner LLC