Principaux documents
Z684732
Ace spherical reaction flask with three vertical necks
capacity 1 L
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
spherical
three-neck (Vertical)
three-neck
Joint
Joints ST/NS24/40 (3)
Description
0-10 mm zero dead space bottom valve
Stérilité
non-sterile
Caractéristiques
cap: no
Conditionnement
case of 1
Fabricant/nom de marque
Ace Glass 6483-110
Capacité
1 L
Description générale
Heavy wall, spherical reaction flask with three vertical necks. Flask has a Zero Dead Space (ZDS) bottom valve. The valve features no material hang-up in the fully open position. When closed, the valve seats flush with the bottom of the flask.