Principaux documents
Z683256
J.Young high-vacuum PTFE valve, right angle
stopcock size 15 mm, side arm O.D. 19 mm
About This Item
Code UNSPSC :
40141600
Nomenclature NACRES :
NB.79
Fabricant/nom de marque
J. Young
D.E.du bras latéral
19 mm
Taille du robinet d'arrêt
15 mm
Description générale
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
  • PTFE piston with unique PTFE-sheathed elastomer O-ring seals
  • Grease-free glass-to-PTFE seals
  • High-vacuum to 10-6 torr