Principaux documents
Z677620
Camatril nitrile long cuff gloves
size XXL
Synonyme(s) :
disposable glove
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Matériaux
green gloves
green nitrile
Taille
XXL
Stérilité
non-sterile
Caractéristiques
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Conditionnement
pack of 1 pair
Fabricant/nom de marque
KCL 733
KCL 733
L × épaisseur
600 mm × 0.5 mm
Description générale
Camatril nitrile long cuff gloves are of high standard, with a wide range of applications and made of specific high quality Nitrile. The large spectrum of chemical resistance ensures the use of these gloves in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials. Good flexibility, anatomical design and excellent fit ensure a good sense of touch.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Roughened palm for firm grip when wet
  • High mechanical resistance
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Informations légales
Camatril is a trademark of KCL GmbH