Principaux documents
Z674273
Duran® GLS 80 caps and accessories
GL 80 blue pouring ring
Synonyme(s) :
1160166
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Fabricant/nom de marque
DURAN® 1160166
Description générale
Permits opening and closing of the DURAN® GLS 80® bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
Caractéristiques et avantages
  • Triple thread permits user friendly opening and closing with only a three-quarter turn
  • Cap offer a linerless, integral lip seal
  • Pouring ring for drip-free pouring and clean, safe working
Informations légales
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group