Principaux documents
Z660477
Julabo® bath test tube trays
holds 360 micro tubes
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.77
Fabricant/nom de marque
Julabo 8960442
Julabo
Disponibilité
available only in EU
Description générale
Carrier tray with 4 racks 8970382 (Z615692-1EA) to hold 360 tubes, tubes sizes 30 x 11/12 mm, suitable for use up to +80 °C.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.