Principaux documents
Z569488
Wilmad® coaxial insert, inner tube
O.D. × I.D. 2.52 mm × 1.5 mm
About This Item
Code UNSPSC :
41121705
Nomenclature NACRES :
SB.32
Fabricant/nom de marque
Wilmad 519-inner tube only
D.E. × D.I.
2.52 mm × 1.5 mm
Description générale
Coaxial insert, inner tube.
Informations légales
Wilmad is a registered trademark of SP Industries