Principaux documents
Z569011
Sigma-Aldrich
Aldrich® Liebig condenser with cup
Overall H 300 mm, joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
H totale
300 mm
Catégories apparentées
Description générale
Condensate cup catches water that forms on the outer surface of the condenser during use and eliminates puddling on reactor and equipment. Tubing connects to cup′s hose barb to drain away water automatically.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC