Principaux documents
Z563129
J-KEM® digital temperature monitor
thermocouple type, T, temp. range -200-250 °C
About This Item
Code UNSPSC :
41112303
Entrée CA/CC
120 V
Caractéristiques
thermocouple type T
Fabricant/nom de marque
J-Kem
Paramètres
-200-250 °C temp. range
Description générale
Displays the temperature of an attached reaction or other piece of equipment on a bright LED display. No alarms. 0.1 °C resolution.
Informations légales
J-KEM is a registered trademark of J-KEM Electronics, Inc.