Principaux documents
Z557595
Sigma-Aldrich
Aldrich® pear-shaped splash-guard adapter
capacity 250 mL, female joint: ST/NS 24/40, male joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS24/40
male joint: ST/NS14/20
Description
anti-climb
Capacité
250 mL
Description générale
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC