Principaux documents
Z552674
Sigma-Aldrich
Aldrich® Thermometer Adapter
straight, male joint: ST/NS 10/18, GL 14 thread
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
male joint: ST/NS10/18
Raccords
GL 14 thread
Catégories apparentées
Description générale
Adapter with 10/18 joint is suited for use with 1/8 in. diameter thermocouple probes.
Has PTFE faced silicone washer and open-top cap.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC