Principaux documents
Z551821
Sigma-Aldrich
Aldrich® Thermometer Adapter
offset, male joint: ST/NS 24/40, GL 14 thread
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
male joint: ST/NS24/40
Raccords
GL 14 thread
Catégories apparentées
Description générale
Has PTFE faced silicone washer and open-top cap.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC