MilliporeSigma
Toutes les photos(6)
Principaux documents
Z544760
Molymod® Individual organic model set
Amino acid models collection set
About This Item
Code UNSPSC :
60104506
Nomenclature NACRES :
NB.17
Description
models 7 (L-config.)
Fabricant/nom de marque
Molymod
Catégories apparentées
Description générale
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Informations légales
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.