Principaux documents
Z532177
Sigma-Aldrich
Aldrich® single bank manifold with glass stopcocks
Manifold with vacuum-gauge port, positions, 4
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.79
Caractéristiques
positions 4
Longueur
400 mm
Description générale
Single bank manifolds come with 4 mm bore glass stopcocks.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC