Principaux documents
Z514918
Kimax® high strength centrifuge tube replacement cap
Synonyme(s) :
73802-24400
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Fabricant/nom de marque
Kimble®
Caractéristiques et avantages
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.