Z510815
Connecting Adapter
female joint: ST/NS 14/20, thread size 20-400
Synonyme(s) :
747130-2014
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Taille du filetage
20-400
Joint
female joint: ST/NS14/20
Catégories apparentées
Description générale
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Caractéristiques et avantages
- Accessory for microscale MICROFLEX threaded kits
- Connects Standard Taper outer ground joint to GPI thread of inner ground joint
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements