Principaux documents
Z402370
Sigma-Aldrich
Best® butyl gloves
size XXL
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Matériaux
black gloves
butyl rubber
Taille
XXL
Stérilité
non-sterile
Caractéristiques
latex free
powder-free
reusable
Conditionnement
pkg of 1 pair
Fabricant/nom de marque
Best Manufacturing 878-11
L × épaisseur
350 mm × 0.70 mm
Caractéristiques et avantages
  • Impermeable
  • Resistant to water
  • Great tactility and performance
  • Provides permeation resistance to gases and water vapors
  • Unsupported, heavy-duty, 100% butyl rubber
  • Injection molded
  • Smooth grip with rolled cuff
  • Offers flexibility and durability
  • Excellent resistance to ketones, acids, caustics, isocyanate, and gases
Informations légales
Best is a registered trademark of Showa Glove Co.