Principaux documents
Z280046
Sigma-Aldrich
Norprene® A-60-G laboratory tubing
I.D. × O.D. 3/4 in. × 1 in.
Synonyme(s) :
Saint Gobain AFL00053, Tygon A-60-G
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 50 feet
Fabricant/nom de marque
Tygon
D.I. × D.E.
3/4 in. × 1 in.
Taille de la paroi
1/8 in.
Catégories apparentées
Description générale
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
  • Chemical resistance to oxidizing agents, acids, and alkalies
  • Resistance to flex-fatigue, will not weaken or crack
Informations légales
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.