Principaux documents
Z244465
Rigid PTFE sleeve
joint: ST/NS 45/50
Synonyme(s) :
ptfe sleeves
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS45/50
Fabricant/nom de marque
Cowie
Description générale
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.