Z219428
Hirsch filter funnel
porosity 25-50 μm (C)
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.43
Produits recommandés
Fabricant/nom de marque
Ace Glass 972703
Diam. du disque
10 mm
Diam. du filtre
10 mm
Dimension de pores
25-50 μm porosity (C)
Capacité
7 mL capacity
Catégories apparentées
Description générale
For use with all microscale flasks and vials with ST/NS: 14/10 joint. Plain stem funnel has a 10 mm diameter sintered disc with a capacity above disc of approximately 7 mL.