Principaux documents
Z207179
Sigma-Aldrich
Broken-glass disposal boxes
Floor
Synonyme(s) :
floor standing waste glass box, laboratory waste glass box
About This Item
Code UNSPSC :
41123300
Nomenclature NACRES :
NB.35
Matériaux
cardboard
Caractéristiques
Holds broken glass
Conditionnement
pkg of 6 ea
Fabricant/nom de marque
Bel-Art F24653-0001
Disponibilité
not available in EU
Larg. × L × H
12 in. × 12 in. × 27 in.
Catégories apparentées
Description générale
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full, safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English, French, Spanish, and German). Shipped flat.