Principaux documents
Z127396
Sigma-Aldrich
Aldrich® large-capacity cold trap
Replacement bulb, for Z106909, with 50 mm O-ring joint
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Capacité du bulbe
500 mL
Catégories apparentées
Description générale
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC