Principaux documents
Z124400
Sigma-Aldrich
Natural rubber septa
red rubber, mixed set (145 pieces with carrying box)
About This Item
Code UNSPSC :
41121800
eCl@ss :
32119211
Nomenclature NACRES :
NB.35
Matériaux
red rubber
Catégories apparentées
Description générale
Mixed set contains:
25 each of the septa for 5 mm NMR tubes; 7 mm O.D. glass tubing; and 8 or 9 mm O.D. glass tubing
20 each of the septa for 9 or 10 mm O.D. glass tubing; and 10/30 joints and small test tubes
10 each of the septa for 14/20 joints; 19/22 joints; and 24/40 joints
Composants de kit également disponibles séparément
Réf. du produit
Description
FDS
  • Z564710Sleeve stopper septa, bottom I.D. × O.D. 9.3 mm × 15.7 mm, red 2 EAFDS
  • Z565644Sleeve stopper septa, bottom I.D. × O.D. 1.5 mm × 3.9 mm, red 5 EAFDS
  • Z565660Sleeve stopper septa, bottom I.D. × O.D. 2.2 mm × 4.5 mm, red 5 EAFDS
  • Z565687Sleeve stopper septa, bottom I.D. × O.D. 2.4 mm × 5.3 mm, red 5 EAFDS
  • Z565709Sleeve stopper septa, bottom I.D. × O.D. 3.4 mm × 6.6 mm, red 4 EAFDS
  • Z564699Sleeve stopper septa, bottom I.D. × O.D. 7.9 mm × 14 mm, red 2 eaFDS
  • Z564699Sleeve stopper septa, bottom I.D. × O.D. 7.9 mm × 14 mm, redFDS
  • Z564737Sleeve stopper septa, bottom I.D. × O.D. 12.7 mm × 23.7 mm, red 2 EAFDS
Afficher tout (8)